Learn More
Abcam Rat monoclonal [SB73a] Anti-Mouse IgM mu chain (Alkaline Phosphatase), 500 uL
Product Code. 32111308 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
Product Code. Quantity Unit Size
32111308 500 μL 500µL
1 options
This item is not returnable. View return policy
Product Code. 32111308 Supplier Abcam Supplier No. ab99589500uL
 Restricted Item
Fecha estimada de envoi: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Rat Monoclonal antibody, suitable for Flow Cyt, ICC/IF, ELISA.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Applications Flow Cytometry
Classification Monoclonal
Clone SB73a
Conjugate Unconjugated
Formulation pH: 8, Preservative: 0.1% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 0.605% Tris, 0.0095% Magnesium chloride
Gene Ighm
Gene Symbols Ighm
Host Species Rat
Purification Method Affinity purification Protein A
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule Ighm, Mouse
Validated Application abrv Flow Cyt, ICC/IF, ELISA