Learn More
Abcam IP10 (CXCL10) Antibody Pair - BSA and Azide free, 10 x 96 Tests
Product Code. 32114003 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32114003 10 x 96 Tests 96 tests
1 options
This item is not returnable. View return policy
Product Code. 32114003 Supplier Abcam Supplier No. ab25384610x96Tests
 Restricted Item
Fecha estimada de envoi: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Antibody, suitable for sELISA.
  • Applications: sELISA
  • Species Reactivity: Human, Rhesus monkey and Cynomolgus monkey
Specifications
Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols CXCL10
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Human, Rhesus monkey and Cynomolgus monkey.
Target Species Human, Rhesus Monkey, Cynomolgus Monkey
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Hu, RhMk
Validated Application abrv sELISA