Learn More
Abcam Human YB1 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Product Code. 32112775 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32112775 10 x 96 Tests 96 tests
1 options
This item is not returnable. View return policy
Product Code. 32112775 Supplier Abcam Supplier No. ab27038110x96Tests
 Restricted Item
Fecha estimada de envoi: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Antibody, suitable for sELISA.
  • Applications: sELISA
  • Species Reactivity: Human
Specifications
Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols YBX1
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Human.
Target Species Human
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv sELISA