missing translation for 'onlineSavingsMsg'
Learn More

A4GALT Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 12392-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16820094

  • 319.00€ / 150 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene catalyzes the transfer of galactose to lactosylceramide to form globotriaosylceramide, which has been identified as the P(k) antigen of the P blood group system. The encoded protein, which is a type II membrane protein found in the Golgi, is also required for the synthesis of the bacterial verotoxins receptor.
Especificaciones

Especificaciones

A4GALT
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q67BJ4, Q9NPC4
A4GALT, A4GALT1, CD77 synthase, Gb3 synthase
Rabbit
IgG
150 μL
-20°C
Primary
239559, 53947
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
A4GALT
Liquid
A4GALT
A4GALT Fusion Protein Ag3069
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documentos
Certificados