Learn More
(9H-Purin-6-ylamino)acetic Acid (may contain up to 20% of inorganics), TRC
Product Code. 30416376
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416376 50 mg 50mg
30412557 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30416376 Supplier Toronto Research Chemicals Supplier No. TRCP84029350MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (9H-Purin-6-ylamino)acetic Acid
CAS 10082-93-6
Molecular Formula C7 H7 N5 O2
Quantity 50 mg
Synonym N-9H-Purin-6-ylglycine, 2-(7H-Purin-6-ylazaniumyl)acetate, 2-[(1H-Purin-6-yl)amino]acetic Acid, 2-[(7H-Purin-6-yl)amino]acetic Acid, 2-[(9H-Purin-6-yl)amino]acetic Acid, NSC 37011
InChI Formula InChI=1S/C7H7N5O2/c13-4(14)1-8-6-5-7(10-2-9-5)12-3-11-6/h2-3H,1H2,(H,13,14)(H2,8,9,10,11,12)
SMILES OC(=O)CNc1ncnc2nc[nH]c12
IUPAC Name 2-(7H-purin-6-ylamino)acetic acid
Molecular Weight (g/mol) 193.163
Formula Weight 193.06