Learn More
Ácido 9-fenilcarbazol-3-borónico, 98 %, Thermo Scientific Chemicals
Catalog Number p-7055469
Click to view available options
Cantidad:
250 mg
1 g
5 g
854952-58-2
C18H14BNO2
287.13
MFCD12196936
JWJQEUDGBZMPAX-UHFFFAOYSA-N
9-phenyl-9h-carbazol-3-yl boronic acid, 9-phenyl-9h-carbazol-3-ylboronic acid, 9-phenyl carbazole-3-boronic acid, 9-phenyl-3-carbazoleboronic acid, 9-phenylcarbazole-3-boronic acid, b-9-phenyl-9h-carbazol-3-yl boronic acid, n-phenylcarbazole-3-boronic acid, 3-borono-9-phenyl-9h-carbazole, pubchem23232, g00061-watson-int
51358450
ácido (9-fenilcarbazol-3-il)borónico
OB(O)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1
854952-58-2
C18H14BNO2
287.13
MFCD12196936
JWJQEUDGBZMPAX-UHFFFAOYSA-N
9-phenyl-9h-carbazol-3-yl boronic acid, 9-phenyl-9h-carbazol-3-ylboronic acid, 9-phenyl carbazole-3-boronic acid, 9-phenyl-3-carbazoleboronic acid, 9-phenylcarbazole-3-boronic acid, b-9-phenyl-9h-carbazol-3-yl boronic acid, n-phenylcarbazole-3-boronic acid, 3-borono-9-phenyl-9h-carbazole, pubchem23232, g00061-watson-int
51358450
ácido (9-fenilcarbazol-3-il)borónico
OB(O)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1
9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El ácido 9-fenilcarbazol-3-borónico se utiliza en la síntesis de colorantes orgánicos funcionalizados.
Solubilidad
Ligeramente soluble en agua.
Notas
Refrigerar. Mantener el recipiente herméticamente cerrado. Almacenar alejado de agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 9-Phenylcarbazole-3-boronic acid
Cantidad 250 mg
Beilstein 10566519
Información de solubilidad Slightly soluble in water.
Formula Weight (peso de la fórmula) 287.13
Porcentaje de pureza 98%
Forma física Sólido