missing translation for 'onlineSavingsMsg'
Learn More

Ácido 9,9-dimetilfluoreno-2-borónico, 98 %, Thermo Scientific™

Catalog Number p-7056038
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
333432-28-3
C15H15BO2
238.093
MFCD08704227
DMDPAJOXRYGXCB-UHFFFAOYSA-N
9,9-dimethyl-9h-fluoren-2-yl boronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-boronic acid, 9,9-dimethylfluoren-2-boronic acid, 9,9-dimethyl-2-fluoreneboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-2-boronic acid, 9,9-dimethylfluorene-2-boronic acid, 9,9-dimethyl-9h-fluorene-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronicacid
22348454
ácido (9,9-dimetilfluoren-2-il)borónico
B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(C)C)(O)O
Este artículo no se puede devolver. Vea la política de devoluciones
333432-28-3
C15H15BO2
238.093
MFCD08704227
DMDPAJOXRYGXCB-UHFFFAOYSA-N
9,9-dimethyl-9h-fluoren-2-yl boronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-boronic acid, 9,9-dimethylfluoren-2-boronic acid, 9,9-dimethyl-2-fluoreneboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-2-boronic acid, 9,9-dimethylfluorene-2-boronic acid, 9,9-dimethyl-9h-fluorene-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronicacid
22348454
ácido (9,9-dimetilfluoren-2-il)borónico
B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(C)C)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 9,9-Dimethylfluorene-2-boronic acid
Cantidad 250 mg
Beilstein 9563230
Formula Weight (peso de la fórmula) 238.09
Porcentaje de pureza 98%
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'