Learn More
8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one, TRC
Product Code. 30351075
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30351075 500 mg 500mg
1 options
This item is not returnable. View return policy
Product Code. 30351075 Supplier Toronto Research Chemicals Supplier No. TRCC365245500MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 8-Chloro-5,6-dihydro-11H-benzo[5.6]cyclohepta[1,2-b]pyridin-11-one
CAS 31251-41-9
Purity Grade Notes HPLC
Molecular Formula C14 H10 Cl N O
Quantity 500 mg
Synonym 8-Chloro-5,6-dihydro-11H-benzo[5.6]cyclohepta[1,2-b]pyridin-11-one, Loratadine Imp. B (EP)
InChI Formula InChI=1S/C14H10ClNO/c15-11-5-6-12-10(8-11)4-3-9-2-1-7-16-13(9)14(12)17/h1-2,5-8H,3-4H2
SMILES Clc1ccc2C(=O)c3ncccc3CCc2c1
IUPAC Name 8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-one
Molecular Weight (g/mol) 243.69
Formula Weight 243.0451
Percent Purity >95