Learn More
8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, Mikromol™
Product Code. 30049221
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049221 25 mg 25mg
17972066 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049221 Supplier Mikromol Supplier No. MM0257.050025
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
CAS 38092-89-6
Analyte or Component Names 8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
Shipping Condition Room Temperature
Molecular Formula C20 H21 Cl N2
Certifications/Compliance ISO 17025
Synonym Loratadine Imp. G (EP), Loratadine USP Related Compound B, Loratadine USP RC B, 8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, Loratadine Related Compound B, Loratadine Impurity G
InChI Formula InChI=1S/C20H21ClN2/c1-23-11-8-14(9-12-23)19-18-7-6-17(21)13-16(18)5-4-15-3-2-10-22-20(15)19/h2-3,6-7,10,13H,4-5,8-9,11-12H2,1H3
SMILES CN1CCC(=C2c3ccc(Cl)cc3CCc4cccnc24)CC1
IUPAC Name 8-chloro-11-(1-methylpiperidin-4-ylidene)-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
Molecular Weight (g/mol) 324.85
Quantity 25 mg
Formula Weight 324.1393 g/mol