Learn More
Integra Biosciences™ 8-Channel Adapter with Ejector
Product Code. 12633685
Change view
Click to view available options
For Use With (Equipment):
VACUSIP Portable Aspirator
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
12633685 VACUSIP Portable Aspirator Each
1 options
This item is not returnable. View return policy
Product Code. 12633685 Supplier Integra Biosciences™ Supplier No. 159024
  • ONLINE EXCLUSIVE PRICE
    54.15€
    51.14€ / paquete de 1
    Ahorro 3.01€
    5% Descuento
View all promotions
Fecha estimada de envoi: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Use this adapter with VACUSIP portable aspirators. Integra Biosciences™ 8-Channel Aspiration Adapters allow VACUSIP aspirators to provide solutions for a wide array of liquid waste disposal tasks.
  • 8-channel adapter with ejector for disposable tips
  • For VACUSIP portable aspirators
  • Unique VACUSIP hand operator allows for fine regulation of the liquid flow and can be connected to various adapters
  • Different adapters are available to match a multitude of laboratory containers
Specifications
Type 8-channel Adapter with Ejector
For Use With (Equipment) VACUSIP Portable Aspirator