Learn More
7-(Trifluoromethyl)1H-indole-2,3-dione, TRC
Product Code. 30451614
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30451614 1 g 1g
1 options
This item is not returnable. View return policy
Product Code. 30451614 Supplier Toronto Research Chemicals Supplier No. TRCT7913001G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 7-(Trifluoromethyl)1H-indole-2,3-dione
CAS 391-12-8
Molecular Formula C9 H4 F3 N O2
Quantity 1 g
InChI Formula InChI=1S/C9H4F3NO2/c10-9(11,12)5-3-1-2-4-6(5)13-8(15)7(4)14/h1-3H,(H,13,14,15)
SMILES FC(F)(F)c1cccc2C(=O)C(=O)Nc12
IUPAC Name 7-(trifluoromethyl)-1H-indole-2,3-dione
Molecular Weight (g/mol) 215.13
Formula Weight 215.0194