missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-2H-1,4-benzotiazina-3(4H)-ona, 97 %, Thermo Scientific™

Catalog Number p-7041101
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
21762-75-4
C8H6N2O3S
210.21
MFCD23379853
OOICZQBVVODNDJ-UHFFFAOYSA-N
7-nitro-2h-benzo b 1,4 thiazin-3 4h-one, 7-nitro-3,4-dihydro-3-oxo-2h 1,4 benzothiazine, 7-nitro-2,4-dihydro-1,4-benzothiazin-3-one, 7-nitro-4h-benzo 1,4 thiazin-3-one, 7-nitro-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 7-nitro, 3,4-dihydro-7-nitro-2h-1,4-benzothiazin-3-one, 7-hydroxy oxido amino-2h-1,4-benzothiazin-3 4h-one
370121
7-nitro-3,4-dihydro-2H-1,4-benzothiazin-3-one
[O-][N+](=O)C1=CC2=C(NC(=O)CS2)C=C1
Este artículo no se puede devolver. Vea la política de devoluciones
21762-75-4
C8H6N2O3S
210.21
MFCD23379853
OOICZQBVVODNDJ-UHFFFAOYSA-N
7-nitro-2h-benzo b 1,4 thiazin-3 4h-one, 7-nitro-3,4-dihydro-3-oxo-2h 1,4 benzothiazine, 7-nitro-2,4-dihydro-1,4-benzothiazin-3-one, 7-nitro-4h-benzo 1,4 thiazin-3-one, 7-nitro-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 7-nitro, 3,4-dihydro-7-nitro-2h-1,4-benzothiazin-3-one, 7-hydroxy oxido amino-2h-1,4-benzothiazin-3 4h-one
370121
7-nitro-3,4-dihydro-2H-1,4-benzothiazin-3-one
[O-][N+](=O)C1=CC2=C(NC(=O)CS2)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 7-Nitro-2H-1,4-benzothiazin-3(4H)-one
Cantidad 250 mg
Formula Weight (peso de la fórmula) 210.21
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'