missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoro-1,2,3,4-tetrahidro-9H-pirido[3,4-b]indol, 95 %, Thermo Scientific™

Catalog Number p-7030224
Click to view available options
Cantidad:
100 mg
500 mg
Este artículo no se puede devolver. Vea la política de devoluciones
177858-80-9
C11H11FN2
190.221
MFCD09264527
OTGGOPSBNVEBDA-UHFFFAOYSA-N
7-fluoro-2,3,4,9-tetrahydro-1h-beta-carboline, 7-fluoro-1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 7-fluoro-1h,2h,3h,4h,9h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, 7-fluoro-1,2,3,4-tetrahydrobeta-carboline, 1h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro
15077550
7-fluoro-2,3,4,9-tetrahidro-1H-pirido[3,4-b]indol
C1CNCC2=C1C3=C(N2)C=C(C=C3)F
Este artículo no se puede devolver. Vea la política de devoluciones
177858-80-9
C11H11FN2
190.221
MFCD09264527
OTGGOPSBNVEBDA-UHFFFAOYSA-N
7-fluoro-2,3,4,9-tetrahydro-1h-beta-carboline, 7-fluoro-1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 7-fluoro-1h,2h,3h,4h,9h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, 7-fluoro-1,2,3,4-tetrahydrobeta-carboline, 1h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro
15077550
7-fluoro-2,3,4,9-tetrahidro-1H-pirido[3,4-b]indol
C1CNCC2=C1C3=C(N2)C=C(C=C3)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 7-Fluoro-1,2,3,4-tetrahydro-9H-pyrido[3,4-b]indole
Punto de fusión 216°C to 224°C
Cantidad 100 mg
Formula Weight (peso de la fórmula) 190.22
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'