missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-5-metilbenzo[b]furano, 97 %, Thermo Scientific Chemicals

Catalog Number p-7032754
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
35700-48-2
C9H7BrO
211.058
MFCD11877828
TZUMDTURHSPEAY-UHFFFAOYSA-N
7-bromo-5-methylbenzofuran, 7-bromo-5-methylbenzo b furan, 7-bromo-5-methyl-benzofuran, benzofuran, 7-bromo-5-methyl
21071801
7-bromo-5-metil-1-benzofurano
CC1=CC(=C2C(=C1)C=CO2)Br
Este artículo no se puede devolver. Vea la política de devoluciones
35700-48-2
C9H7BrO
211.058
MFCD11877828
TZUMDTURHSPEAY-UHFFFAOYSA-N
7-bromo-5-methylbenzofuran, 7-bromo-5-methylbenzo b furan, 7-bromo-5-methyl-benzofuran, benzofuran, 7-bromo-5-methyl
21071801
7-bromo-5-metil-1-benzofurano
CC1=CC(=C2C(=C1)C=CO2)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 7-Bromo-5-methylbenzo[b]furan
Cantidad 250 mg
Formula Weight (peso de la fórmula) 211.06
Porcentaje de pureza 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'