missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1,2,3,4-tetrahidrociclopent[b]indol, 99 %, Thermo Scientific™

Catalog Number p-7033472
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
164736-47-4
C11H10BrN
236.112
MFCD00239009
XKDWSCYYHKJAGK-UHFFFAOYSA-N
7-bromo-1,2,3,4-tetrahydrocyclopenta b indole, 7-bromo-1,2,3,4-tetrahydrocyclopent b indole, 7-bromo-1h,2h,3h,4h-cyclopenta b indole, cyclopent b indole, 7-bromo-1,2,3,4-tetrahydro, 7-bromo-1,2,3-trihydrocyclopenta 1,2-b indole, bio6e6, acmc-20a4tv
700805
7-bromo-1,2,3,4-tetrahidrociclopenta[b]indol
C1CC2=C(C1)NC3=C2C=C(C=C3)Br
Este artículo no se puede devolver. Vea la política de devoluciones
164736-47-4
C11H10BrN
236.112
MFCD00239009
XKDWSCYYHKJAGK-UHFFFAOYSA-N
7-bromo-1,2,3,4-tetrahydrocyclopenta b indole, 7-bromo-1,2,3,4-tetrahydrocyclopent b indole, 7-bromo-1h,2h,3h,4h-cyclopenta b indole, cyclopent b indole, 7-bromo-1,2,3,4-tetrahydro, 7-bromo-1,2,3-trihydrocyclopenta 1,2-b indole, bio6e6, acmc-20a4tv
700805
7-bromo-1,2,3,4-tetrahidrociclopenta[b]indol
C1CC2=C(C1)NC3=C2C=C(C=C3)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole
Cantidad 250 mg
Formula Weight (peso de la fórmula) 236.11
Porcentaje de pureza 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'