Learn More
7-(2-Carbomethoxyethyl)-5-(2-chlorophenyl)-thieno-1,4-diazepin-2-one, TRC
Product Code. 30367101
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30367101 10 mg 10mg
1 options
This item is not returnable. View return policy
Product Code. 30367101 Supplier Toronto Research Chemicals Supplier No. TRCC17662010MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 7-(2-Carbomethoxyethyl)-5-(2-chlorophenyl)-thieno-1,4-diazepin-2-one
CAS 100827-80-3
Molecular Formula C17 H15 Cl N2 O3 S
Quantity 10 mg
InChI Formula InChI=1S/C17H15ClN2O3S/c1-23-15(22)7-6-10-8-12-16(11-4-2-3-5-13(11)18)19-9-14(21)20-17(12)24-10/h2-5,8H,6-7,9H2,1H3,(H,20,21)
SMILES COC(=O)CCc1cc2C(=NCC(=O)Nc2s1)c3ccccc3Cl
IUPAC Name methyl 3-[5-(2-chlorophenyl)-2-oxo-1,3-dihydrothieno[2,3-e][1,4]diazepin-7-yl]propanoate
Molecular Weight (g/mol) 362.83
Formula Weight 362.0492