Learn More
6-Hydroxy Bromantane, TRC
Product Code. 30008999
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008999 25 mg 25mg
30004410 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008999 Supplier Toronto Research Chemicals Supplier No. TRCH83001525MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1007310-57-7
Molecular Formula C16H20BrNO
Molecular Weight (g/mol) 322.24
Synonym Tricyclo[3.3.1.13,7]decan-2-ol, 6-[(4-bromophenyl)amino]-, 6-[(4-Bromophenyl)amino]tricyclo[3.3.1.13,7]decan-2-ol, 6-Hydroxybromantan
SMILES OC1[C@@H]2C[C@@H]3C[C@H]1C[C@H](C2)C3Nc4ccc(Br)cc4
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C16H20BrNO/c17-13-1-3-14(4-2-13)18-15-9-5-11-7-10(15)8-12(6-9)16(11)19/h1-4,9-12,15-16,18-19H,5-8H2
Formula Weight 321.0728
Percent Purity >95%
Chemical Name or Material 6-Hydroxy Bromantane
Grade HPLC