missing translation for 'onlineSavingsMsg'
Learn More

6-Fluorocromona, 97 %, Thermo Scientific Chemicals

Catalog Number p-7030992
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
105300-38-7
C9H5FO2
164.135
MFCD03094002
WHIGSYZUTMYUAX-UHFFFAOYSA-N
6-fluorochromone, 6-fluoro-4h-chromen-4-one, 6-fluoro-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-fluoro, acmc-20ebjx, 6-fluoranylchromen-4-one, 6-fluoro-1-benzopyran-4-one, 6-fluoro-4-oxo-4h-1-benzopyran
688892
6-fluorocromo-4-ona
C1=CC2=C(C=C1F)C(=O)C=CO2
Este artículo no se puede devolver. Vea la política de devoluciones
105300-38-7
C9H5FO2
164.135
MFCD03094002
WHIGSYZUTMYUAX-UHFFFAOYSA-N
6-fluorochromone, 6-fluoro-4h-chromen-4-one, 6-fluoro-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-fluoro, acmc-20ebjx, 6-fluoranylchromen-4-one, 6-fluoro-1-benzopyran-4-one, 6-fluoro-4-oxo-4h-1-benzopyran
688892
6-fluorocromo-4-ona
C1=CC2=C(C=C1F)C(=O)C=CO2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 6-Fluorochromone
Punto de fusión 166°C to 169°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 164.14
Porcentaje de pureza 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'