missing translation for 'onlineSavingsMsg'
Learn More

6-Cianoimidazo[1,2-a]piridina, 95 %, Thermo Scientific Chemicals

Catalog Number p-7032962
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
106850-34-4
C8H5N3
143.149
MFCD06659603
LRJOKNYELSECDZ-UHFFFAOYSA-N
imidazo 1,2-a pyridine-6-carbonitrile, 6-cyanoimidazo 1,2-a pyridine, 4-hydroimidazo 1,2-a pyridine-6-carbonitrile, 6-cyano-imidazo 1,2-a pyridine, acmc-1bu34, 6-imidazo 1,2-a pyridinecarbonitrile, imidazo 1,2-, a pyridine-6-carbonitrile, imidazo 1,2-alpha pyridine-6-carbonitrile, 6-cyanoimidazo 1,2-a pyridine-6-carbonitrile
2764471
imidazo[1,2-a]piridina-6-carbonitrilo
C1=CC2=NC=CN2C=C1C#N
Este artículo no se puede devolver. Vea la política de devoluciones
106850-34-4
C8H5N3
143.149
MFCD06659603
LRJOKNYELSECDZ-UHFFFAOYSA-N
imidazo 1,2-a pyridine-6-carbonitrile, 6-cyanoimidazo 1,2-a pyridine, 4-hydroimidazo 1,2-a pyridine-6-carbonitrile, 6-cyano-imidazo 1,2-a pyridine, acmc-1bu34, 6-imidazo 1,2-a pyridinecarbonitrile, imidazo 1,2-, a pyridine-6-carbonitrile, imidazo 1,2-alpha pyridine-6-carbonitrile, 6-cyanoimidazo 1,2-a pyridine-6-carbonitrile
2764471
imidazo[1,2-a]piridina-6-carbonitrilo
C1=CC2=NC=CN2C=C1C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 6-Cyanoimidazo[1,2-a]pyridine
Punto de fusión 161°C to 163°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 148.15
Porcentaje de pureza 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'