missing translation for 'onlineSavingsMsg'
Learn More

6-Clorotiocroman-4-ona, 98 %, Thermo Scientific Chemicals

Catalog Number p-7080673
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
13735-12-1
C9H7ClOS
198.664
MFCD00052997
OKHUUKHZUNKSQA-UHFFFAOYSA-N
6-chlorothiochroman-4-one, 6-chloro-2,3-dihydro-4h-thiochromen-4-one, 6-chloro-3,4-dihydro-2h-1-benzothiopyran-4-one, 6-chloro-thiochroman-4-one, 6-chloro-3,4-dihydro-2h-1-benzothiin-4-one, 6-chloro-2,3-dihydro-4h-1-benzothiopyran-4-one, 6-chloro-2,3-dihydro-1-benzothiopyran-4-one, 6-chloro-2h,3h-benzo e thiin-4-one, acmc-20alz1, maybridge1_007431
248065
6-cloro-2,3-dihidrotiocromen-4-ona
C1CSC2=C(C1=O)C=C(C=C2)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
13735-12-1
C9H7ClOS
198.664
MFCD00052997
OKHUUKHZUNKSQA-UHFFFAOYSA-N
6-chlorothiochroman-4-one, 6-chloro-2,3-dihydro-4h-thiochromen-4-one, 6-chloro-3,4-dihydro-2h-1-benzothiopyran-4-one, 6-chloro-thiochroman-4-one, 6-chloro-3,4-dihydro-2h-1-benzothiin-4-one, 6-chloro-2,3-dihydro-4h-1-benzothiopyran-4-one, 6-chloro-2,3-dihydro-1-benzothiopyran-4-one, 6-chloro-2h,3h-benzo e thiin-4-one, acmc-20alz1, maybridge1_007431
248065
6-cloro-2,3-dihidrotiocromen-4-ona
C1CSC2=C(C1=O)C=C(C=C2)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 6-Chlorothiochroman-4-one
Punto de fusión 65°C to 69°C
Cantidad 1 g
Beilstein 124621
Formula Weight (peso de la fórmula) 198.67
Porcentaje de pureza 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'