Learn More
6-Chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic Acid, Mikromol™
Product Code. 30051188
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051188 25 mg 25mg
30050316 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051188 Supplier Mikromol Supplier No. MM0080.030025
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-Chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic Acid
CAS 88190-77-6
Analyte or Component Names 6-Chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C15 H11 Cl N2 O2
Certifications/Compliance ISO 9001
Synonym 6-Chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic Acid
InChI Formula InChI=1S/C15H11ClN2O2/c16-10-6-7-12-11(8-10)13(9-4-2-1-3-5-9)18-14(17-12)15(19)20/h1-8,13H,(H,17,18)(H,19,20)
SMILES OC(=O)C1=Nc2ccc(Cl)cc2C(N1)c3ccccc3
IUPAC Name 6-chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic acid
Molecular Weight (g/mol) 286.71
Quantity 25 mg
Formula Weight 286.0509 g/mol