missing translation for 'onlineSavingsMsg'
Learn More

6''-Cloro-3,2':5',3''-terpiridina, 95 %

Catalog Number p-7030037
Click to view available options
Cantidad:
100 mg
500 mg
Este artículo no se puede devolver. Vea la política de devoluciones
942206-13-5
C15H10ClN3
267.716
MFCD09037442
HFAGORVNAKFNGR-UHFFFAOYSA-N
6'-chloro-3,2':5',3'-terpyridine, 5-6-chloropyridin-3-yl-2-pyridin-3-yl pyridine, 6-chloro-3,2':5',3-terpyridine, 6-chloro-3,2':5',3' terpyridine, 3∼6∼-chloro-1∼3∼,2∼2∼:2∼5∼,3∼3∼-terpyridine, 6-chloro-6'-pyridin-3-yl-3,3'-bipyridine
42553044
2-cloro-5-(6-piridin-3-ilpiridin-3-il)piridina
C1=CC(=CN=C1)C2=NC=C(C=C2)C3=CN=C(C=C3)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
942206-13-5
C15H10ClN3
267.716
MFCD09037442
HFAGORVNAKFNGR-UHFFFAOYSA-N
6'-chloro-3,2':5',3'-terpyridine, 5-6-chloropyridin-3-yl-2-pyridin-3-yl pyridine, 6-chloro-3,2':5',3-terpyridine, 6-chloro-3,2':5',3' terpyridine, 3∼6∼-chloro-1∼3∼,2∼2∼:2∼5∼,3∼3∼-terpyridine, 6-chloro-6'-pyridin-3-yl-3,3'-bipyridine
42553044
2-cloro-5-(6-piridin-3-ilpiridin-3-il)piridina
C1=CC(=CN=C1)C2=NC=C(C=C2)C3=CN=C(C=C3)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 6''-Chloro-3,2':5',3''-terpyridine
Cantidad 100 mg
Formula Weight (peso de la fórmula) 267.72
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'