missing translation for 'onlineSavingsMsg'
Learn More

(6-Bromohexiloxi)-tert-butildimetilsilano, 99 %, Thermo Scientific Chemicals

Catalog Number p-7052739
Click to view available options
Cantidad:
5 mL
25 mL
Este artículo no se puede devolver. Vea la política de devoluciones
129368-70-3
C12H27BrOSi
295.336
MFCD01863708
PBKXRKYUUXKNSL-UHFFFAOYSA-N
6-bromohexyloxy-tert-butyldimethylsilane, 6-bromohexyl oxy tert-butyl dimethylsilane, silane, 6-bromohexyl oxy 1,1-dimethylethyl dimethyl, acmc-20apme, 1-t-butyldimethylsilyloxy-6-bromohexane, 6-bromohexyloxy-tert-butyidimethylsilane, 6-bromohexyloxy tert-butyldimethylsilane, 6-bromohexyl tert-butyldimethylsilyl ether
4260352
6-bromohexoxi-tert-butil-dimetilsilano
CC(C)(C)[Si](C)(C)OCCCCCCBr
Este artículo no se puede devolver. Vea la política de devoluciones
129368-70-3
C12H27BrOSi
295.336
MFCD01863708
PBKXRKYUUXKNSL-UHFFFAOYSA-N
6-bromohexyloxy-tert-butyldimethylsilane, 6-bromohexyl oxy tert-butyl dimethylsilane, silane, 6-bromohexyl oxy 1,1-dimethylethyl dimethyl, acmc-20apme, 1-t-butyldimethylsilyloxy-6-bromohexane, 6-bromohexyloxy-tert-butyidimethylsilane, 6-bromohexyloxy tert-butyldimethylsilane, 6-bromohexyl tert-butyldimethylsilyl ether
4260352
6-bromohexoxi-tert-butil-dimetilsilano
CC(C)(C)[Si](C)(C)OCCCCCCBr

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Punto de ebullición 276°C
Nombre del producto químico o material (6-Bromohexyloxy)-tert-butyldimethylsilane
Color Incolora
Forma física Líquido
Cantidad 5 mL
Temperatura de inflamación 110°C (230°F)
Formula Weight (peso de la fórmula) 295.34
Porcentaje de pureza 99%
Índice de refracción 1.457
Densidad 1.053 g/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'