Learn More
6,7-Dimethoxyquinazoline-2,4-dione, TRC
Product Code. 30377450
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377450 10 g 10g
30380874 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30377450 Supplier Toronto Research Chemicals Supplier No. TRCD47495010G
 Restricted Item
Fecha estimada de envoi: 16-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28888-44-0
Molecular Formula C10 H10 N2 O4
Molecular Weight (g/mol) 222.2
Synonym 6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione, Doxazosin Mesilate Imp. D (EP), 2,4(1H,3H)-Quinazolinedione, 6,7-dimethoxy-, 6,7-Dimethoxy-2,4(1H,3H)-quinazolinedione, 2,5-Dihydroxy-6,7-dimethoxyquinazoline, 6,7-Dimethoxy-1H-quinazoline-2,4-dione, 6,7-Dimethoxyquinazoline-2,4-diol, 6,7-Dimethoxyquinazoline-2,4-dione
IUPAC Name 6,7-dimethoxy-1H-quinazoline-2,4-dione
SMILES COc1cc2NC(=O)NC(=O)c2cc1OC
Specifications
Quantity 10 g
InChI Formula InChI=1S/C10H10N2O4/c1-15-7-3-5-6(4-8(7)16-2)11-10(14)12-9(5)13/h3-4H,1-2H3,(H2,11,12,13,14)
Formula Weight 222.0641
Chemical Name or Material 6,7-dimethoxyquinazoline-2,4(1H,3H)-dione