missing translation for 'onlineSavingsMsg'
Learn More

Cloruro de 5-(fenilsulfonil)tiofeno-2-sulfonilo, téc. 90 %

Catalog Number p-7045716
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
166964-37-0
C10H7ClO4S3
322.792
MFCD00052143
GSLVRDAYMHUGTF-UHFFFAOYSA-N
5-phenylsulfonyl thiophene-2-sulfonyl chloride, 5-phenylsulfonyl thiophene-2-sulfonylchloride, 5-benzenesulfonyl thiophene-2-sulfonyl chloride, 2-benzenesulfonylthiophene-5-sulfonyl chloride, 5-phenylthiophene-2,5-disulphonyl chloride, pubchem5455, 5-benzenesulphonylthiophene-2-sulphonyl chloride, acmc-1c42s, 5-phenylsulphonyl thiophene-2-sulphonyl chloride, 2-phenylsulfonylthiophene-5-sulfonyl chloride
580877
cloruro de 5-(bencenosulfonil)tiofeno-2-sulfonilo
C1=CC=C(C=C1)S(=O)(=O)C2=CC=C(S2)S(=O)(=O)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
166964-37-0
C10H7ClO4S3
322.792
MFCD00052143
GSLVRDAYMHUGTF-UHFFFAOYSA-N
5-phenylsulfonyl thiophene-2-sulfonyl chloride, 5-phenylsulfonyl thiophene-2-sulfonylchloride, 5-benzenesulfonyl thiophene-2-sulfonyl chloride, 2-benzenesulfonylthiophene-5-sulfonyl chloride, 5-phenylthiophene-2,5-disulphonyl chloride, pubchem5455, 5-benzenesulphonylthiophene-2-sulphonyl chloride, acmc-1c42s, 5-phenylsulphonyl thiophene-2-sulphonyl chloride, 2-phenylsulfonylthiophene-5-sulfonyl chloride
580877
cloruro de 5-(bencenosulfonil)tiofeno-2-sulfonilo
C1=CC=C(C=C1)S(=O)(=O)C2=CC=C(S2)S(=O)(=O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-(Phenylsulfonyl)thiophene-2-sulfonyl chloride
Punto de fusión 102°C to 105°C
Cantidad 250 mg
Número UN UN3261
Beilstein 7818655
Sensibilidad Moisture sensitive
Formula Weight (peso de la fórmula) 322.81
Porcentaje de pureza 90%
Grado Técnico
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'