Learn More
5-Methylnicotinamide, TRC
Product Code. 30339470
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30339470 25 mg 25mg
1 options
This item is not returnable. View return policy
Product Code. 30339470 Supplier Toronto Research Chemicals Supplier No. TRCB49530825MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Methylnicotinamide
CAS 70-57-5
Molecular Formula C7H8N2O
Quantity 25 mg
Synonym NSC 87668, 5-Methyl-3-pyridinecarboxamide
InChI Formula InChI=1S/C7H8N2O/c1-5-2-6(7(8)10)4-9-3-5/h2-4H,1H3,(H2,8,10)
SMILES Cc1cncc(c1)C(=O)N
IUPAC Name 5-methylpyridine-3-carboxamide
Molecular Weight (g/mol) 136.15
Formula Weight 136.0637