missing translation for 'onlineSavingsMsg'
Learn More

Ácido 5-metil-3-fenilisoxazol-4-carboxílico, 99 %, Thermo Scientific Chemicals

Catalog Number p-7043525
Click to view available options
Cantidad:
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
1136-45-4
C11H9NO3
203.197
MFCD00003153
PENHKTNQUJMHIR-UHFFFAOYSA-N
5-methyl-3-phenylisoxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-4-isoxazolecarboxylic acid, 3-phenyl-5-methylisoxazol-4-carbonsaeure, 3-phenyl-5-methylisoxazole-4-carboxylic acid, isoxazole-4-carboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-isoxazole-4-carboxylic acid, 5-methyl-3-phenylisoxazole-4-carboxylicacid, 3-phenyl-5-methylisoxazol-4-carbonsaeure german, 5-methyl-3-phenyl-4-isooxazolic acid
14343
ácido 5-metil-3-fenil-1,2-oxazol-4-carboxílico
CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)O
Este artículo no se puede devolver. Vea la política de devoluciones
1136-45-4
C11H9NO3
203.197
MFCD00003153
PENHKTNQUJMHIR-UHFFFAOYSA-N
5-methyl-3-phenylisoxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-4-isoxazolecarboxylic acid, 3-phenyl-5-methylisoxazol-4-carbonsaeure, 3-phenyl-5-methylisoxazole-4-carboxylic acid, isoxazole-4-carboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-isoxazole-4-carboxylic acid, 5-methyl-3-phenylisoxazole-4-carboxylicacid, 3-phenyl-5-methylisoxazol-4-carbonsaeure german, 5-methyl-3-phenyl-4-isooxazolic acid
14343
ácido 5-metil-3-fenil-1,2-oxazol-4-carboxílico
CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-Methyl-3-phenylisoxazole-4-carboxylic acid
Punto de fusión 191°C to 193°C
Olor Odorless
Cantidad 25 g
Beilstein 164939
Formula Weight (peso de la fórmula) 203.2
Porcentaje de pureza 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'