missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-phenyl-3-furoic acid, 97%, Thermo Scientific™

Product Code. 10123184
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10123184 5 g 5g
10717495 250 mg 250mg
10491884 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10123184 Supplier Thermo Scientific Maybridge Supplier No. CC49701DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 64354-50-3
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD00447900
InChI Key CASZQQKUMVLKBY-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-3-furoic acid, 3-furancarboxylic acid,5-methyl-2-phenyl, 5-methyl-2-phenyl-furan-3-carboxylic acid, cbmicro_037588, phenuvic acid, cambridge id 5926078
PubChem CID 761020
IUPAC Name 5-methyl-2-phenylfuran-3-carboxylic acid
SMILES CC1=CC(=C(O1)C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-phenyl-3-furoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.