Learn More
5-Hydroxymethyl-N-phenyl-2-1H-pyridone-d5, TRC
Product Code. 30398495
Change view
Click to view available options
Quantity:
1 mg
Product Code. Quantity
30398495 1 mg
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30398495 Supplier Toronto Research Chemicals Supplier No. TRCH9482771MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Hydroxymethyl-N-phenyl-2-1H-pyridone-d5
CAS 1020719-52-1
Molecular Formula C12 2H5 H6 N O2
Quantity 1 mg
Synonym 2(1H)-Pyridinone, 5-(hydroxymethyl)-1-(phenyl-2,3,4,5,6-d5)-, 5-(Hydroxymethyl)-1-(phenyl-d5)-2(1H)-pyridinone, 5-(Hydroxymethyl)-1-phenyl-2(1H)-pyridinone-D5 (phenyl-D5), 5-Hydroxymethyl-N-phenyl-2-1H-pyridone-d5
InChI Formula InChI=1S/C12H11NO2/c14-9-10-6-7-12(15)13(8-10)11-4-2-1-3-5-11/h1-8,14H,9H2/i1D,2D,3D,4D,5D
SMILES [2H]c1c([2H])c([2H])c(N2C=C(CO)C=CC2=O)c([2H])c1[2H]
IUPAC Name 5-(hydroxymethyl)-1-(2,3,4,5,6-pentadeuteriophenyl)pyridin-2-one
Molecular Weight (g/mol) 206.25
Formula Weight 206.1104