missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-3-(3-oxiciclopentil)indol, 95 %, Thermo Scientific™

Catalog Number p-7055374
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
675831-63-7
C13H12FNO
217.243
DINNCXVUOXLHSH-UHFFFAOYSA-N
3-5-fluoro-1h-indol-3-yl cyclopentanone, 3-5fluoro-1h-indol-3-yl-cyclopentanone, 3-5-fluoro-1h-indol-3-yl-cyclopentanone, 3-5-fluoro-1h-indol-3-yl cyclopentan-1-one
22139466
3-(5-fluoro-1H-indol-3-il)ciclopentan-1-ona
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)F
Este artículo no se puede devolver. Vea la política de devoluciones
675831-63-7
C13H12FNO
217.243
DINNCXVUOXLHSH-UHFFFAOYSA-N
3-5-fluoro-1h-indol-3-yl cyclopentanone, 3-5fluoro-1h-indol-3-yl-cyclopentanone, 3-5-fluoro-1h-indol-3-yl-cyclopentanone, 3-5-fluoro-1h-indol-3-yl cyclopentan-1-one
22139466
3-(5-fluoro-1H-indol-3-il)ciclopentan-1-ona
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-Fluoro-3-(3-oxycyclopentyl)indole
Cantidad 250 mg
Formula Weight (peso de la fórmula) 217.24
Porcentaje de pureza 95%
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'