Learn More
5-Fluoro-2-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
Product Code. 11476356
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476356 1 g 1g
11486356 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476356 Supplier Thermo Scientific Alfa Aesar Supplier No. B23135.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 239135-48-9
Molecular Formula C8H5BrF4
Molecular Weight (g/mol) 257.03
MDL Number MFCD00236290
InChI Key MQZWBTRGSDOAIO-UHFFFAOYSA-N
Synonym 5-fluoro-2-trifluoromethyl benzyl bromide, 2-bromomethyl-4-fluoro-1-trifluoromethyl benzene, 2-trifluoromethyl-5-fluorobenzyl bromide, acmc-1cfdc, 5-fluoro-2-trifluoromethylbenzyl bromide, 2-trifluoromethyl-5-fluoro-benzyl bromide, alpha-bromo-5-fluoro-2-trifluoromethyl toluene, benzene,2-bromomethyl-4-fluoro-1-trifluoromethyl, a-bromo-5-fluoro-2-trifluoromethyl toluene;a-bromo-5-fluoro-2-trifluoromethyl toluene
PubChem CID 2737578
IUPAC Name 2-(bromomethyl)-4-fluoro-1-(trifluoromethyl)benzene
SMILES FC1=CC(CBr)=C(C=C1)C(F)(F)F
Specifications
Refractive Index 1.4817
Quantity 1 g
UN Number UN1760
Sensitivity Moisture sensitive
Formula Weight 257.03
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)benzyl bromide