missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-metilanisol, 98 %, Thermo Scientific Chemicals

Catalog Number p-7028359
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
95729-22-9
C8H9FO
140.157
MFCD00673008
CXTIUQIZWCABPV-UHFFFAOYSA-N
5-fluoro-2-methylanisole, 4-fluoro-2-methoxytoluene, 4-fluoro-2-methoxy-1-methyl-benzene, pubchem2624, 1-methyl-2-methoxy-4-fluorobenzene, 5-fluoro-2-methylphenyl methyl ether, 4-fluoranyl-2-methoxy-1-methyl-benzene, benzene, 4-fluoro-2-methoxy-1-methyl
2774573
4-fluoro-2-metoxi-1-metilbenceno
CC1=C(C=C(C=C1)F)OC
Este artículo no se puede devolver. Vea la política de devoluciones
95729-22-9
C8H9FO
140.157
MFCD00673008
CXTIUQIZWCABPV-UHFFFAOYSA-N
5-fluoro-2-methylanisole, 4-fluoro-2-methoxytoluene, 4-fluoro-2-methoxy-1-methyl-benzene, pubchem2624, 1-methyl-2-methoxy-4-fluorobenzene, 5-fluoro-2-methylphenyl methyl ether, 4-fluoranyl-2-methoxy-1-methyl-benzene, benzene, 4-fluoro-2-methoxy-1-methyl
2774573
4-fluoro-2-metoxi-1-metilbenceno
CC1=C(C=C(C=C1)F)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-Fluoro-2-methylanisole
Índice de refracción 1.494
Cantidad 1 g
Formula Weight (peso de la fórmula) 140.16
Porcentaje de pureza 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'