missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzo[b]tiofeno, + 98 %, Thermo Scientific Chemicals

Catalog Number p-7048036
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
4923-87-9
C8H5BrS
213.09
MFCD03069318
RDSIMGKJEYNNLF-UHFFFAOYSA-N
5-bromobenzo b thiophene, 5-bromobenzothiophene, 5-bromo-benzo b thiophene, 5-bromothianaphthene, benzo b thiophene, 5-bromo, zlchem 181, pubchem7752, 5-bromo-benzothiophene, pubchem15353, acmc-1anox
2776578
5-bromo-1-benzotiofeno
BrC1=CC=C2SC=CC2=C1
Este artículo no se puede devolver. Vea la política de devoluciones
4923-87-9
C8H5BrS
213.09
MFCD03069318
RDSIMGKJEYNNLF-UHFFFAOYSA-N
5-bromobenzo b thiophene, 5-bromobenzothiophene, 5-bromo-benzo b thiophene, 5-bromothianaphthene, benzo b thiophene, 5-bromo, zlchem 181, pubchem7752, 5-bromo-benzothiophene, pubchem15353, acmc-1anox
2776578
5-bromo-1-benzotiofeno
BrC1=CC=C2SC=CC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-Bromobenzo[b]thiophene
Punto de fusión 44°C to 48°C
Punto de ebullición 105°C to 107°C (10 mmHg)
Cantidad 250 mg
Beilstein 112878
Sensibilidad Light sensitive
Formula Weight (peso de la fórmula) 213.1
Porcentaje de pureza ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'