missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-metoxipirimidina, 95 %, Thermo Scientific™

Catalog Number p-7030999
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
14001-66-2
C5H5BrN2O
189.012
MFCD04973351
DWVCZDMMGYIULX-UHFFFAOYSA-N
2-methoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2-methoxy, 5-bromo-2-methoxy-pyrimidine, 5-bromo-2-methoxy-1,3-diazine, 5-bromopyrimidin-2-yl methyl ether, 5-bromo-2-methyloxy pyrimidine, pubchem5264, acmc-209cjt, 5-bromo-2methoxypyrimidine, ksc494g8n
139657
5-bromo-2-metoxipirimidina
COC1=NC=C(C=N1)Br
Este artículo no se puede devolver. Vea la política de devoluciones
14001-66-2
C5H5BrN2O
189.012
MFCD04973351
DWVCZDMMGYIULX-UHFFFAOYSA-N
2-methoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2-methoxy, 5-bromo-2-methoxy-pyrimidine, 5-bromo-2-methoxy-1,3-diazine, 5-bromopyrimidin-2-yl methyl ether, 5-bromo-2-methyloxy pyrimidine, pubchem5264, acmc-209cjt, 5-bromo-2methoxypyrimidine, ksc494g8n
139657
5-bromo-2-metoxipirimidina
COC1=NC=C(C=N1)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-Bromo-2-methoxypyrimidine
Cantidad 250 mg
Formula Weight (peso de la fórmula) 189.01
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'