missing translation for 'onlineSavingsMsg'
Learn More

Ácido 5-bromo-2-metoxicinámico, 98 %, Thermo Scientific™

Catalog Number 11443388
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
40803-53-0
C10H8BrO3
256.08
MFCD00017596
QABZTGNRWBRJBO-GORDUTHDSA-M
5-bromo-2-methoxycinnamic acid, 3-5-bromo-2-methoxyphenyl acrylic acid, 2e-3-5-bromo-2-methoxyphenyl prop-2-enoic acid, 2e-3-5-bromo-2-methoxyphenyl acrylic acid, e-3-5-bromo-2-methoxyphenyl acrylic acid, substance h 47, 3-bromo-6-methoxycinnamic acid, acide bromo-5-methoxy-2-cinnamique, trans-3-bromo-6-methoxycinnamic acid, 3-5-bromo-2-methoxyphenyl propenoic acid
676162
(2E)-3-(5-bromo-2-methoxyphenyl)prop-2-enoate
COC1=CC=C(Br)C=C1\C=C\C([O-])=O
Este artículo no se puede devolver. Vea la política de devoluciones
40803-53-0
C10H8BrO3
256.08
MFCD00017596
QABZTGNRWBRJBO-GORDUTHDSA-M
5-bromo-2-methoxycinnamic acid, 3-5-bromo-2-methoxyphenyl acrylic acid, 2e-3-5-bromo-2-methoxyphenyl prop-2-enoic acid, 2e-3-5-bromo-2-methoxyphenyl acrylic acid, e-3-5-bromo-2-methoxyphenyl acrylic acid, substance h 47, 3-bromo-6-methoxycinnamic acid, acide bromo-5-methoxy-2-cinnamique, trans-3-bromo-6-methoxycinnamic acid, 3-5-bromo-2-methoxyphenyl propenoic acid
676162
(2E)-3-(5-bromo-2-methoxyphenyl)prop-2-enoate
COC1=CC=C(Br)C=C1\C=C\C([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-Bromo-2-methoxycinnamic acid
Punto de fusión 224°C to 226°C
Cantidad 25 g
Beilstein 3265594
Formula Weight (peso de la fórmula) 257.09
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'