missing translation for 'onlineSavingsMsg'
Learn More

Alcohol 5-bromo-2-hidroxibencílico, + 98 %, Thermo Scientific™

Catalog Number p-7082078
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
2316-64-5
C7H7BrO2
203.04
MFCD00004618
KNKRHSVKIORZQB-UHFFFAOYSA-N
5-bromo-2-hydroxybenzyl alcohol, 4-bromo-2-hydroxymethyl phenol, bromosaligenin, 5-bromosalicyl alcohol, bromsalizol, 5-bromosaligenin, 5-bromo-2-hydroxybenzylalcohol, 2-hydroxymethyl-4-bromophenol, benzenemethanol, 5-bromo-2-hydroxy, unii-y1uz53815e
75342
4-bromo-2-(hidroximetil)fenol
OCC1=CC(Br)=CC=C1O
Este artículo no se puede devolver. Vea la política de devoluciones
2316-64-5
C7H7BrO2
203.04
MFCD00004618
KNKRHSVKIORZQB-UHFFFAOYSA-N
5-bromo-2-hydroxybenzyl alcohol, 4-bromo-2-hydroxymethyl phenol, bromosaligenin, 5-bromosalicyl alcohol, bromsalizol, 5-bromosaligenin, 5-bromo-2-hydroxybenzylalcohol, 2-hydroxymethyl-4-bromophenol, benzenemethanol, 5-bromo-2-hydroxy, unii-y1uz53815e
75342
4-bromo-2-(hidroximetil)fenol
OCC1=CC(Br)=CC=C1O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-Bromo-2-hydroxybenzyl alcohol
Punto de fusión 108°C to 110°C
Cantidad 5 g
Beilstein 509726
Formula Weight (peso de la fórmula) 203.04
Porcentaje de pureza ≥98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'