Learn More
5-Amino-4,6-dicloropirimidina, +98 %, Thermo Scientific Chemicals
Catalog Number p-7027919
Click to view available options
Cantidad:
1 g
5 g
25 g
5413-85-4
C4H3Cl2N3
163.99
MFCD00006108
NIGDWBHWHVHOAD-UHFFFAOYSA-N
5-amino-4,6-dichloropyrimidine, 5-pyrimidinamine, 4,6-dichloro, 4,6-dichloro-5-pyrimidinamine, 4,6-dichloro-5-aminopyrimidine, 4,6-dichloropyrimidin-5-ylamine, 5-amine-4,6-dichloropyrimidine, 5-amino-4,6-dichloropyfimidine, 4,6-dichloro-pyrimidin-5-ylamine, pyrimidine, 5-amino-4,6-dichloro, 4,6-dichloropyrimidine-5-ylamine
79434
4,6-dicloropirimidín-5-amina
NC1=C(Cl)N=CN=C1Cl
5413-85-4
C4H3Cl2N3
163.99
MFCD00006108
NIGDWBHWHVHOAD-UHFFFAOYSA-N
5-amino-4,6-dichloropyrimidine, 5-pyrimidinamine, 4,6-dichloro, 4,6-dichloro-5-pyrimidinamine, 4,6-dichloro-5-aminopyrimidine, 4,6-dichloropyrimidin-5-ylamine, 5-amine-4,6-dichloropyrimidine, 5-amino-4,6-dichloropyfimidine, 4,6-dichloro-pyrimidin-5-ylamine, pyrimidine, 5-amino-4,6-dichloro, 4,6-dichloropyrimidine-5-ylamine
79434
4,6-dicloropirimidín-5-amina
NC1=C(Cl)N=CN=C1Cl
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-Amino-4,6-dichloropyrimidine
Punto de fusión 143°C to 146°C
Olor Irritating
Cantidad 1 g
Beilstein 126885
Sensibilidad Moisture sensitive
Formula Weight (peso de la fórmula) 164
Porcentaje de pureza ≥98%