missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-(4-clorofenil)-1H-pirazol, 97 %, Thermo Scientific™

Catalog Number p-7031124
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
78583-81-0
C9H8ClN3
193.63
MFCD00053046
XQPBZIITFQHIDI-UHFFFAOYSA-N
3-4-chlorophenyl-1h-pyrazol-5-amine, 5-4-chlorophenyl-1h-pyrazol-3-amine, 5-amino-3-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-amine, 3-amino-5-4-chlorophenyl-1h-pyrazole, 5-amino-3-4-chlorophenyl-1h-pyrazole, 3-amino-5-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-ylamine, 3-4-chlorophenyl pyrazole-5-ylamine, pubchem12126
2735305
5-(4-chlorophenyl)-1H-pyrazol-3-amine
NC1=NNC(=C1)C1=CC=C(Cl)C=C1
Este artículo no se puede devolver. Vea la política de devoluciones
78583-81-0
C9H8ClN3
193.63
MFCD00053046
XQPBZIITFQHIDI-UHFFFAOYSA-N
3-4-chlorophenyl-1h-pyrazol-5-amine, 5-4-chlorophenyl-1h-pyrazol-3-amine, 5-amino-3-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-amine, 3-amino-5-4-chlorophenyl-1h-pyrazole, 5-amino-3-4-chlorophenyl-1h-pyrazole, 3-amino-5-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-ylamine, 3-4-chlorophenyl pyrazole-5-ylamine, pubchem12126
2735305
5-(4-chlorophenyl)-1H-pyrazol-3-amine
NC1=NNC(=C1)C1=CC=C(Cl)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-Amino-3-(4-chlorophenyl)-1H-pyrazole
Punto de fusión 172°C to 174°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 193.64
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'