Learn More
5-Amino-1-metil-3-fenil-1H-pirazol, 97 %, Thermo Scientific Chemicals
Catalog Number p-7022826
Click to view available options
Cantidad:
1 g
5 g
25 g
10199-50-5
C10H11N3
173.219
MFCD00067874
KCYRMURRLLYLPU-UHFFFAOYSA-N
1-methyl-3-phenyl-1h-pyrazol-5-amine, 5-amino-1-methyl-3-phenylpyrazole, 5-amino-1-methyl-3-phenyl-1h-pyrazole, 2-methyl-5-phenyl-2h-pyrazol-3-ylamine, 1-methyl-3-phenylpyrazole-5-ylamine, acmc-1bxci, maybridge1_004465, 1-methyl-3-phenyl-5-pyrazolamine, 1-methyl-3-phenyl-pyrazol-5-amine, 2-methyl-5-phenyl-pyrazol-3-amine
517779
2-metil-5-fenilpirazol-3-amina
CN1C(=CC(=N1)C2=CC=CC=C2)N
10199-50-5
C10H11N3
173.219
MFCD00067874
KCYRMURRLLYLPU-UHFFFAOYSA-N
1-methyl-3-phenyl-1h-pyrazol-5-amine, 5-amino-1-methyl-3-phenylpyrazole, 5-amino-1-methyl-3-phenyl-1h-pyrazole, 2-methyl-5-phenyl-2h-pyrazol-3-ylamine, 1-methyl-3-phenylpyrazole-5-ylamine, acmc-1bxci, maybridge1_004465, 1-methyl-3-phenyl-5-pyrazolamine, 1-methyl-3-phenyl-pyrazol-5-amine, 2-methyl-5-phenyl-pyrazol-3-amine
517779
2-metil-5-fenilpirazol-3-amina
CN1C(=CC(=N1)C2=CC=CC=C2)N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-Amino-1-methyl-3-phenyl-1H-pyrazole
Punto de fusión 124°C to 130°C
Olor Odorless
Cantidad 1 g
Beilstein 743807
Formula Weight (peso de la fórmula) 173.22
Porcentaje de pureza 97%