Learn More
5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid, TRC
Product Code. 30413157
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30413157 5 g 5g
1 options
This item is not returnable. View return policy
Product Code. 30413157 Supplier Toronto Research Chemicals Supplier No. TRCT2941005G
 Restricted Item
Fecha estimada de envoi: 22-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid
CAS 4242-18-6
Molecular Formula C11 H12 O2
Quantity 5 g
Synonym 1-Naphthalenecarboxylic acid','5,6,7,8-tetrahydro-, 1-Naphthoic acid','5,6,7,8-tetrahydro- (6CI,7CI,8CI), 5,6,7,8-Tetrahydro-1-naphthalenecarboxylic acid, 5,6,7,8-Tetrahydro-1-naphthoic acid, 5,6,7,8-Tetrahydronaphthalen-1-carboxylic acid, NSC 44874, 5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid
InChI Formula InChI=1S/C11H12O2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h3,5,7H,1-2,4,6H2,(H,12,13)
SMILES OC(=O)c1cccc2CCCCc12
IUPAC Name 5,6,7,8-tetrahydronaphthalene-1-carboxylic acid
Molecular Weight (g/mol) 176.21
Formula Weight 176.0837