missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8,9,10-Hexahidrociclohept[b]indol, 98 %, Thermo Scientific Chemicals

Catalog Number p-7024527
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
2047-89-4
C13H15N
185.27
MFCD00101340
XZUJMYLNFZHNLP-UHFFFAOYSA-N
5,6,7,8,9,10-hexahydrocyclohepta b indole, 5,6,7,8,9,10-hexahydro-cyclohepta b indole, 5h,6h,7h,8h,9h,10h-cyclohepta b indole, cycloheptan a indole, 2,3-pentano-1h-indole, 5,6,7,8,9,10-hexahydrocyclohept b indole, 5,6,7,8,9,10-hexahydrocyclohepta b indole #, 6,7,8,9,10,10a-hexahydrocyclohepta b indole, 6,7,8,9,10-pentahydrocyclohepta 1,2-b indole, 6,7,8,9,10-pentahydrocyclohepta 2,1-b indole
251955
5,6,7,8,9,10-Hexahidrociclohepta[b]indol
C1CCC2=C(CC1)NC3=CC=CC=C23
Este artículo no se puede devolver. Vea la política de devoluciones
2047-89-4
C13H15N
185.27
MFCD00101340
XZUJMYLNFZHNLP-UHFFFAOYSA-N
5,6,7,8,9,10-hexahydrocyclohepta b indole, 5,6,7,8,9,10-hexahydro-cyclohepta b indole, 5h,6h,7h,8h,9h,10h-cyclohepta b indole, cycloheptan a indole, 2,3-pentano-1h-indole, 5,6,7,8,9,10-hexahydrocyclohept b indole, 5,6,7,8,9,10-hexahydrocyclohepta b indole #, 6,7,8,9,10,10a-hexahydrocyclohepta b indole, 6,7,8,9,10-pentahydrocyclohepta 1,2-b indole, 6,7,8,9,10-pentahydrocyclohepta 2,1-b indole
251955
5,6,7,8,9,10-Hexahidrociclohepta[b]indol
C1CCC2=C(CC1)NC3=CC=CC=C23

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5,6,7,8,9,10-Hexahydrocyclohept[b]indole
Punto de fusión 143°C to 146°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 185.27
Porcentaje de pureza 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'