missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Metoxifenil)tiofeno-2-carboxaldehído, 99 %, Thermo Scientific Chemicals

Catalog Number p-7033379
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
38401-67-1
C13H12O2S
232.30
MFCD00663639
LRUBIGCXVSIWPI-UHFFFAOYSA-N
5-4-methoxyphenyl thiophene-2-carbaldehyde, 5-4-methoxyphenyl thiophene-2-carboxaldehyde, 5-4-methoxyphenyl-2-thiophenecarbaldehyde, acmc-1ahp9, rarechem ak ma k025, methoxyphenylthiophenecarbaldehyde, 2-formyl-5-4-methoxyphenyl thiophene, 5-4-methoxylphenyl thiophene-2-carbaldehyde, 5-4-methoxyphenyl-2-thiophenecarboxaldehyde, 5-4-methoxy-phenyl-thiophene-2-carbaldehyde
2797800
5-(4-metoxifenil)tiofeno-2-carbaldehído
COC1=CC=C(C=C1)C1=CC=C(S1)C(C)=O
Este artículo no se puede devolver. Vea la política de devoluciones
38401-67-1
C13H12O2S
232.30
MFCD00663639
LRUBIGCXVSIWPI-UHFFFAOYSA-N
5-4-methoxyphenyl thiophene-2-carbaldehyde, 5-4-methoxyphenyl thiophene-2-carboxaldehyde, 5-4-methoxyphenyl-2-thiophenecarbaldehyde, acmc-1ahp9, rarechem ak ma k025, methoxyphenylthiophenecarbaldehyde, 2-formyl-5-4-methoxyphenyl thiophene, 5-4-methoxylphenyl thiophene-2-carbaldehyde, 5-4-methoxyphenyl-2-thiophenecarboxaldehyde, 5-4-methoxy-phenyl-thiophene-2-carbaldehyde
2797800
5-(4-metoxifenil)tiofeno-2-carbaldehído
COC1=CC=C(C=C1)C1=CC=C(S1)C(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 5-(4-Methoxyphenyl)thiophene-2-carboxaldehyde
Cantidad 1 g
Formula Weight (peso de la fórmula) 218.28
Porcentaje de pureza 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'