Learn More
5-(4-Bromophenyl)-4,6-dichloropyrimidine, TRC
Product Code. 30340888
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity Unit Size
30340888 25 g 25g
1 options
This item is not returnable. View return policy
Product Code. 30340888 Supplier Toronto Research Chemicals Supplier No. TRCB68786525G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-(4-Bromophenyl)-4,6-dichloropyrimidine
CAS 146533-41-7
Molecular Formula C10 H5 Br Cl2 N2
Quantity 25 g
Synonym 4,6-Dichloro-5-(4-bromophenyl)pyrimidine
InChI Formula InChI=1S/C10H5BrCl2N2/c11-7-3-1-6(2-4-7)8-9(12)14-5-15-10(8)13/h1-5H
SMILES Clc1ncnc(Cl)c1c2ccc(Br)cc2
IUPAC Name 5-(4-bromophenyl)-4,6-dichloropyrimidine
Molecular Weight (g/mol) 303.97
Formula Weight 301.901