Learn More
Fisherbrand™ 4D Multifunctional Armrests
Product Code. 17766689 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Adjustable:
Adjustable in height, width, depth and swivelling
Unit Size:
Each
Product Code. Adjustable Unit Size
17766689 Adjustable in height, width, depth and swivelling Each
1 options
Product Code. 17766689
  • ONLINE EXCLUSIVE PRICE
    125.00€
    93.77€ / paquete de 1
    Ahorro 31.23€
    24% Descuento
Fecha estimada de envoi: 14-12-2026
Add to basket
Edge
Add to basket
Compatible with Basic Comfort and Active Comfort Lab chairs to relieve the strain on the upper body and arms, especially for fine motor activities.
  • Ergonomic
  • Lab chair accessory
Specifications
Product Type Armrest
Color Black
Adjustable Adjustable in height, width, depth and swivelling
For Use With (Equipment) Fisherbrand Basic Comfort and Active Comfort Lab Chairs
Frame Material Polyamide
Warranty 10 years
Weight (Metric) 3 kg
Width (Metric) 8 cm