missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)aniline hydrochloride, 97%

Product Code. 11413897
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413897 25 g 25g
11403897 5 g 5g
11423897 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413897 Supplier Thermo Scientific Alfa Aesar Supplier No. A18329.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 90774-69-9 | C7H7ClF3N | 197.585 g/mol

4-(Trifluoromethyl)aniline hydrochloride is used in the synthesis of 2-(4-((6-Chloro-2-quinoxalinyl)oxy)phenoxy)-N-(4-(trifluoromethyl)phenyl)propanamide. It is also used to produce N-(p-trifluoromethylphenyl)phosphoramidic dichloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)aniline hydrochloride is used in the synthesis of 2-(4-((6-Chloro-2-quinoxalinyl)oxy)phenoxy)-N-(4-(trifluoromethyl)phenyl)propanamide. It is also used to produce N-(p-trifluoromethylphenyl)phosphoramidic dichloride.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90774-69-9
Molecular Formula C7H7ClF3N
Molecular Weight (g/mol) 197.585
MDL Number MFCD00007897
InChI Key CZGDYZDUMJDQSD-UHFFFAOYSA-N
Synonym 4-aminobenzotrifluoride hydrochloride, 4-trifluoromethyl aniline hydrochloride, 4-trifluoromethyl anilinium chloride, p-trifluoromethylaniline hydrochloride, 4-trifluoromethyl phenylamine, chloride, pubchem2816, 4-aminobenzotrifluoride, hydrochloride, 4-trifluoromethylaniline hydrochloride, 4-trifluoromethyl aniline;hydrochloride, a,a,a-trifluoro-p-toluidine hydrochloride
PubChem CID 2723961
IUPAC Name 4-(trifluoromethyl)aniline;hydrochloride
SMILES C1=CC(=CC=C1C(F)(F)F)N.Cl

Specifications

Melting Point ∼240°C (decomposition)
Quantity 25 g
UN Number UN2811
Beilstein 3629991
Formula Weight 197.59
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)aniline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.