missing translation for 'onlineSavingsMsg'
Learn More

Ácido 4-(trifluorometoxi)benzoico, 97 %, Thermo Scientific™

Catalog Number p-7021332
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
330-12-1
C8H5F3O3
206.12
MFCD00002541
RATSANVPHHXDCT-UHFFFAOYSA-N
4-trifluoromethoxy benzoic acid, p-trifluoromethoxybenzoic acid, 4-trifluoromethoxy-benzoic acid, p-carboxyphenyl trifluoromethyl ether, 4-trifluoromethoxy benzoicacid, benzoic acid, 4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-p-anisic acid, pubchem4593, acmc-1cp95, ksc225q9l
67613
ácido 4-(trifluorometoxi)benzoico
C1=CC(=CC=C1C(=O)O)OC(F)(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
330-12-1
C8H5F3O3
206.12
MFCD00002541
RATSANVPHHXDCT-UHFFFAOYSA-N
4-trifluoromethoxy benzoic acid, p-trifluoromethoxybenzoic acid, 4-trifluoromethoxy-benzoic acid, p-carboxyphenyl trifluoromethyl ether, 4-trifluoromethoxy benzoicacid, benzoic acid, 4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-p-anisic acid, pubchem4593, acmc-1cp95, ksc225q9l
67613
ácido 4-(trifluorometoxi)benzoico
C1=CC(=CC=C1C(=O)O)OC(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-(Trifluoromethoxy)benzoic acid
Punto de fusión 151°C to 154°C
Cantidad 1 g
Beilstein 977356
Formula Weight (peso de la fórmula) 206.12
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'