missing translation for 'onlineSavingsMsg'
Learn More

4-terc-Butoxibenzonitrilo, 96 %, Thermo Scientific™

Catalog Number p-7047218
missing translation for 'orderingAttributeHoverText'
Cantidad:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
185259-36-3
C11H13NO
175.231
MFCD03425273
WLWYCCYGVDZFMS-UHFFFAOYSA-N
4-tert-butoxybenzonitrile, 4-tert-butoxy benzonitrile, benzonitrile, 4-1,1-dimethylethoxy, acmc-20amau, 4-tert-butoxy benzenecarbonitrile, 4-2-methylpropan-2-yl oxy benzonitrile
7021978
4-[(2-metilpropan-2-il)oxi]benzonitrilo
CC(C)(C)OC1=CC=C(C=C1)C#N
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
185259-36-3
C11H13NO
175.231
MFCD03425273
WLWYCCYGVDZFMS-UHFFFAOYSA-N
4-tert-butoxybenzonitrile, 4-tert-butoxy benzonitrile, benzonitrile, 4-1,1-dimethylethoxy, acmc-20amau, 4-tert-butoxy benzenecarbonitrile, 4-2-methylpropan-2-yl oxy benzonitrile
7021978
4-[(2-metilpropan-2-il)oxi]benzonitrilo
CC(C)(C)OC1=CC=C(C=C1)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-tert-Butoxybenzonitrile
Punto de ebullición 95°C (0.8 mmHg)
Índice de refracción 1.521
Cantidad 5 g
Beilstein 3237723
Formula Weight (peso de la fórmula) 175.23
Porcentaje de pureza 96%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'