missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoato de nitrofenilo, 97 %, Thermo Scientific Chemicals

Catalog Number p-7046726
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
959-22-8
C13H9NO4
243.218
MFCD00135494
GMKZBFFLCONHDE-UHFFFAOYSA-N
p-nitrophenyl benzoate, benzoic acid, 4-nitrophenyl ester, benzoic acid, p-nitrophenyl ester, 4-nitrophenyl benzoate, phenol, p-nitro-, benzoate, benzoic acid 4-nitrophenyl ester, benzoic acid,4-nitrophenyl ester
70396
benzoato (4-nitrofenil)
C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-]
Este artículo no se puede devolver. Vea la política de devoluciones
959-22-8
C13H9NO4
243.218
MFCD00135494
GMKZBFFLCONHDE-UHFFFAOYSA-N
p-nitrophenyl benzoate, benzoic acid, 4-nitrophenyl ester, benzoic acid, p-nitrophenyl ester, 4-nitrophenyl benzoate, phenol, p-nitro-, benzoate, benzoic acid 4-nitrophenyl ester, benzoic acid,4-nitrophenyl ester
70396
benzoato (4-nitrofenil)
C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Nitrophenyl benzoate
Punto de fusión 142°C to 144°C
Cantidad 5 g
Beilstein 2054709
Formula Weight (peso de la fórmula) 243.22
Porcentaje de pureza 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'