missing translation for 'onlineSavingsMsg'
Learn More

4'-[N-Etil-N-(2-metilfenil)sulfamoil]acetanilida, 97 %, Thermo Scientific™

Catalog Number p-7040981
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
333430-43-6
C17H20N2O3S
332.42
MFCD01213966
NGMYZQIEURPXLI-UHFFFAOYSA-N
n-4-ethyl 2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-o-tolyl-sulfamoyl-phenyl-acetamide, 4'-n-ethyl-n-2-methylphenyl sulfamoyl acetanilide, n-4-ethyl-2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-2-methylanilino sulfonyl phenyl acetamide, n-4-ethyl 2-methylphenyl amino sulfonyl phenyl acetamide
743585
N-[4-[etil-(2-metilfenil)sulfamoil]fenil]acetamida
CCN(C1=CC=CC=C1C)S(=O)(=O)C1=CC=C(NC(C)=O)C=C1
Este artículo no se puede devolver. Vea la política de devoluciones
333430-43-6
C17H20N2O3S
332.42
MFCD01213966
NGMYZQIEURPXLI-UHFFFAOYSA-N
n-4-ethyl 2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-o-tolyl-sulfamoyl-phenyl-acetamide, 4'-n-ethyl-n-2-methylphenyl sulfamoyl acetanilide, n-4-ethyl-2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-2-methylanilino sulfonyl phenyl acetamide, n-4-ethyl 2-methylphenyl amino sulfonyl phenyl acetamide
743585
N-[4-[etil-(2-metilfenil)sulfamoil]fenil]acetamida
CCN(C1=CC=CC=C1C)S(=O)(=O)C1=CC=C(NC(C)=O)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4'-[N-Ethyl-N-(2-methylphenyl)sulfamoyl]acetanilide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 332.42
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'