Learn More
4-Methylumbelliferyl 2-Acetamido-2-deoxy-Beta-D-galactopyranoside, TRC
Product Code. 30483894
Change view
Click to view available options
Quantity:
25 mg
500 mg
Unit Size:
25mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483894 500 mg 500mg
30471631 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30483894 Supplier Toronto Research Chemicals Supplier No. TRCM333530500MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 36476-29-6
Molecular Formula C18H21NO8
Molecular Weight (g/mol) 379.36
Synonym 4-Methylumbelliferyl beta-D-Acetylgalactosaminide, 4-Methylumbelliferyl N-Acetyl-beta-D-galactosaminide, GalNAc1-b-4MU
IUPAC Name N-[(2S,3R,4R,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]acetamide
SMILES CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CO)O[C@H]1Oc2ccc3C(=CC(=O)Oc3c2)C
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C18H21NO8/c1-8-5-14(22)26-12-6-10(3-4-11(8)12)25-18-15(19-9(2)21)17(24)16(23)13(7-20)27-18/h3-6,13,15-18,20,23-24H,7H2,1-2H3,(H,19,21)/t13-,15-,16+,17-,18-/m1/s1
Formula Weight 379.1267
Chemical Name or Material 4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-galactopyranoside